Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NISTRM8573

L-Glutamic acid

NIST®RM 8573, USGS40

Synonym(s):

(S)-2-Aminopentanedioic acid, Glu

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
147.13
Beilstein/REAXYS Number:
1723801
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

mp

205 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1

InChI key

WHUUTDBJXJRKMK-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biochem/physiol Actions

An excitatory amino acid neurotransmitter that is an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA).

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service