NISTRM8395
Tissue engineering reference scaffold
NIST® RM 8395
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
grade
certified reference material
manufacturer/tradename
NIST®
application(s)
pharmaceutical (small molecule)
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
Storage Class
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service