Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NISTRM8096

CMOS MEMS 5-in-1 test chip

NIST® RM 8096

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

chips

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service