Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST971

Hormones in Frozen Human Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

description

NIST SRM 971

packaging

pkg of 2 x 5 mL

manufacturer/tradename

NIST®

application(s)

clinical

format

matrix material

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service