Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST927E

Bovine Serum Albumin (7% solution)

NIST® SRM® 927e

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 10 x 2.2 mL

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

single component solution

General description

This Standard Reference Material (SRM) is intended primarily for use in the standardization of procedures employed in clinical analyses for total serum protein, for critical evaluation of daily working standards used in these procedures, and as a reference standard for assays of total protein by colorimetric methods. This SRM is a solution (mass fraction 7 %) of known protein concentration and purity. A unit of SRM 927e consists of 10 ampoules, each containing approximately 2.2 mL of solution. For more information, please refer to the SDS and COA.

SRM 927E _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinming Jia et al.
Experimental and therapeutic medicine, 15(3), 2929-2935 (2018-03-31)
The transplantation of neural stem cells (NSCs) has been demonstrated as a potential treatment strategy for traumatic brain injury (TBI). Cu, Zn-superoxide dismutase (SOD1) is an important antioxidant enzyme that detoxifies intracellular reactive oxygen species, thereby protecting cells from oxidative
Yanhui Li et al.
Stem cell research & therapy, 8(1), 156-156 (2017-07-06)
Smooth muscle progenitor cells (pSMCs) differentiated from human pluripotent stem cells (hPSCs) hold great promise for treating diseases or degenerative conditions involving smooth muscle pathologies. However, the therapeutic potential of pSMCs derived from men and women may be very different.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service