Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST781D2

Molybdenum - heat capacity

NIST® SRM® 781D2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

rod

packaging

pkg of 10 cm

manufacturer/tradename

NIST®

General description

This Standard Reference Material (SRM) is intended primarily for high-temperature calorimetric standard. A unit of SRM 781-D2 consists of a rod 0.64 cm (0.25 inches) in diameter by 10 cm (4 inches) in length. For more information, please refer to the COA and SDS.

SRM 781D2_cert SRM 781D2 _SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Molybdenum (Mo)

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service