Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3532

Calcium-containing solid oral dosage form

NIST® SRM® 3532

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 5 x 10 g

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for validation of methods for determining cholecalciferol (vitamin D3) and elements in a calcium dietary supplement and similar materials. This SRM can also be used for quality assurance when assigning values to in-house reference materials. A unit of SRM 3532 consists of five packets, each containing approximately 10 g of material. For more information, please refer to the SDS and COA.

SRM 3532_cert

SRM 3532_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service