Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST331A

Copper ore

NIST® SRM® 331a, mill tails

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

form

powder

packaging

pkg of 40 g

Looking for similar products? Visit Product Comparison Guide

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Carc. 1B - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service