Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3280

Multivitamin/multielement tablets

NIST® SRM® 3280

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 30 tablets x 5 bottles

manufacturer/tradename

NIST®

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in validating analytical methods for the determination of vitamins, carotenoids, and elements in dietary supplement tablets and similar matrices. This SRM can also be used for quality assurance when assigning values to in-house control materials. A unit of SRM 3280 consists of five bottles, each containing 30 tablets. Each tablet weighs approximately 1.5 g. For more information, please refer to the COA and SDS.

SRM 3280_cert

SRM 3280_SDS

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phosphorus (P)
Trace Elements: Antimony (SB), Arsenic (As), Boron (B), Cadmium (Cd), Calcium (Ca), Chloride, Chromium (Cr), Cobalt (Co), Copper (Cu), Iodine (I), Iron (Fe), Lanthanum (La), Lead (Pb), Magnesium (Mg), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Potassium (K), Selenium (Se), Silicon (Si), Sodium (Na), Strontium (Sr), Tin (Sn), Titanium (Ti), Vanadium (V), Zinc (Zn)
Vitamins: Ascorbic acid (Vitamin C), Biotin, cis-β-Carotene, Ergocalciferol, Folic acid, Lutein, Niacinamide (Vitamin B3), Pantothenic acid (Vitamin B5), Pyridoxine hydrochloride, Retinol, Riboflavin (Vitamin B2), Thiamine hydrochloride, Total β-Carotene, trans-β-Carotene, Vitamin B12, Vitamin K1, α-Tocopherol

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service