NIST3117A
Europium (Eu) standard solution
NIST® SRM® 3117a
About This Item
Recommended Products
grade
certified reference material
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
application(s)
pharmaceutical (small molecule)
format
single component solution
General description
SRM 3117A_cert SRM 3117A _SDS
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
supp_hazards
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service