Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3103A

Arsenic (As) standard solution

NIST® SRM® 3103a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

form

solution

packaging

pkg of 5 x 10 mL

application(s)

pharmaceutical (small molecule)

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of arsenic. The solution contains nitric acid at a volume fraction of approximately 10 %, which is equivalent to a concentration (molarity) of approximately 1.6 mol/L. For more information, please refer to the SDS and COA.

SRM 3103A_cert

SRM 3103A_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service