Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2976

Mussel tissue (trace elements and methylmercury), freeze-dried

NIST® SRM® 2976

Synonym(s):

Freeze-dried Mussel Tissue for Trace Element and Methylmercury Analysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 25 g

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

related product

Product No.
Description
Pricing

hcodes

Hazard Classifications

Aquatic Chronic 3

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service