Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2842

Semiconductor thin film: AlxGa1-xAs epitaxial layers (Al mole fraction x near 0.30)

NIST® SRM® 2842

Sign Into View Organizational & Contract Pricing

Select a Size

1 DISC
$2,900.00

$2,900.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 DISC
$2,900.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,900.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

film

packaging

pkg of disk

manufacturer/tradename

NIST®

application(s)

semiconductor

General description

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Aluminum (Al)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service