Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2670A

Toxic elements in freeze-dried urine

NIST® SRM® 2670a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

Quality Level

form

solid

packaging

pkg of set (4) (2 ea conc)

application(s)

pharmaceutical (small molecule)

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Speciation of arsenic in biological samples.
Mandal K B, et al.
Toxicology and Applied Pharmacology, 198(3), 307-318 (2004)
Elimination of the argon chloride interference on arsenic speciation in inductively coupled plasma mass spectrometry using ion chromatography.
Sheppard S B, et al.
Journal of Analytical Atomic Spectrometry, 5(6), 431-435 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service