Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2627A

Nitric oxide in nitrogen

NIST® SRM® 2627a, nominal 5 umol/mol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200

grade

certified reference material

packaging

pkg of cylinder

application(s)

pharmaceutical (small molecule)

format

mixture

InChI

1S/HNO/c1-2/h1H

InChI key

ODUCDPQEXGNKDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

pictograms

Gas cylinder

signalword

Warning

hcodes

pcodes

Hazard Classifications

Press. Gas Liquefied gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service