NIST2614A
Carbon monoxide in air
NIST® SRM® 2614a, nominal 45 μmol/mol
About This Item
grade
certified reference material
packaging
pkg of cylinder
manufacturer/tradename
NIST®
application(s)
pharmaceutical (small molecule)
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
SRM 2614A_cert
SRM 2614A_SDS
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service