Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2112

Dynamic impact force verification specimens

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of set

manufacturer/tradename

NIST®

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for the verification of the maximum force measured dynamically using an instrumented Charpy impact machine in accordance with the current ASTM Standard E2298 or ISO Standard14556. A unit of SRM 2112 consists of a set of four specimens needed to perform a single verification. SRM 2112 can also be used to verify the absorbed energy scale of the impact machine at the high energy level. For more information,please refer to the SDS and COA.

SRM 2112_cert

SRM 2112_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service