Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST19H

Basic electric steel

NIST® SRM® 19h, 0.2% carbon

Sign Into View Organizational & Contract Pricing

Select a Size

150 G
$902.00

$902.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

Select a Size

Change View
150 G
$902.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$902.00


Available to ship onMay 01, 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 150 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use primarily in chemical methods of analysis. For more information, please refer to the SDS and COA.

SRM 19H_cert

SRM 19H_SDS

Other Notes

Certified for the analytes listed below.
Carbon (C), Manganese (Mn), Phosphorus (P), Sulfur (S), Silicon (Si), Copper (Cu), Nickel (Ni), Chromium (Cr), Vanadium (V), Molybdenum (Mo), Aluminium (Al)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service