NIST1976B
Instrument response standard for x-ray powder diffraction
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
packaging
pkg of 1 disc
application(s)
pharmaceutical (small molecule)
General description
This Standard Reference Material (SRM) consists of a sintered alumina disc intended for use in calibration of X-ray powder diffraction equipment with respect to line position and intensity as a function of 2? angle. The solid form of the SRM eliminates any variability in intensity measurements introduced by the sample loading procedure. A unit of SRM 1976b consists of a sintered alumina disc approximately 25.6 mm in diameter by 2.2 mm in thickness. For more information, please refer to the SDS and COA.
SRM 1976B_cert SRM 1976B _SDS
SRM 1976B_cert SRM 1976B _SDS
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service