Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1955

Homocysteine and folate in frozen human serum

NIST® SRM® 1955

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

form

liquid

packaging

pkg of set (3) (1 ea conc)

application(s)

pharmaceutical (small molecule)

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service