NIST1849A
Infant/adult nutritional formula
NIST® SRM® 1849a
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 10 pouches x 10 g ea
manufacturer/tradename
NIST®
application(s)
cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)
format
matrix material
General description
SRM 1849A_cert
SRM 1849A_SDS
Other Notes
Solids, Ash, Protein, Carbohydrates, Lactose Monohydrate, Calories, Fat (extracted), Fat (as the sum of fatty acids as triglycerides), Saturated Fatty acids, Cis-Monounsaturated Fatty acids, Cis-Polyunsaturated acids, Omega-3 Fatty acids, Omega-6 Fatty acids, Total trans C18:1 Fatty acids, Total trans Fatty acids, Cholesterol, Phosphorous (P)
Amino acids: Alanine, Arginine, Aspartic acid, Cysteine, Glutamic acid, Glycine, Histidine, Isoleucine, Leucine, Lysine, Phenylalanine, Proline, Serine, Taurine, Threonine, Tryptophan, Tyrosine, Valine
Lipids and Fatty acids: Caproic acid, Capric acid, Caprylic acid, Lauric acid, Myristic acid, Margaric acid, Palmitic acid, Palmitoleic acid, Stearic acid, Oleic acid, Vaccenic acid, Linoleic acid, a-Linoleic acid, Arachidic acid, Arachidonic acid, Behenic acid, DHA, Lignoceric acid, Nervonic acid, (Z,E)-9,12-Octadecadienoic acid (C18:2), (E,Z)-9,12-Octadecadienoic acid (C18:2), (Z,Z,Z)-6,9,12-Octadecatrienoic acid (C18:3 n-6), Gondoic acid
Trace Elements: Calcium (Ca), Copper (Cu), Chromium (Cr), Iodine (I), Magnesium (Mg), Manganese (Mn), Molybdenum (Mo), Potassium (K), Selenium (Se), Sodium (Na), Zinc Zn)
Vitamins: Ascorbic acid (Vitamin C), Biotin, Carnitine, Cholecalciferol (Vitamin D3), Choline Ion, Folic acid, Niacinamide (Vitamin B3), Pantothenic acid (Vitamin B5), Pyridoxine (Vitamin B6), Retinol (Vitamin A), Retinyl Palmitate (Vitamin A), Riboflavin (Vitamin B2), Thiamine (Vitamin B1), α-Tocopheryl acetate (Vitamin E), Total α-Tocopherol (Vitamin E), Phylloquinone (Vitamin K1), 5-Methyltetrahydrofolate, myo-Inositol
Nucleotides: Adenosine Monophosphate, Cytidine Monophosphate, Guanosine Monophosphate, Uridine Monophosphate
Matrix Group: Dairy products and eggs
Legal Information
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service