Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1762A

Low alloy steel

NIST® SRM® 1762a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a low alloy steel. This SRM is intended for use in the evaluation of chemical and instrumental methods of analysis and in calibration of analyses. A unit of SRM 1762a consists of a disk approximately 34 mm in diameter and 19 mm thick. For more information, please refer to the COA and SDS.

SRM 1762A_cert SRM 1762A _SDS

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Niobium (Nb), Arsenic (As), Phosphorus (P), Boron (B), Silicon (Si), Carbon (C), Sulfur (S), Cobalt (Co), Tantalum (Ta), Chromium (Cr), Tin (Sn), Copper (Cu), Titanium (Ti), Manganese (Mn), Vanadium (V), Molybdenum (Mo), Zirconium (Zr), Nickel (Ni)

Reference/Informational value is provided for the following analytes. Iron (Fe), Nitrogen (N), Tungsten (W)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service