Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1655

Potassium chloride

NIST® SRM® 1655, KCl (cr) for solution calorimetry

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

powder

packaging

pkg of 30 g

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Thermal Analysis and Calorimetry: Recent Advances, Techniques and Applications (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service