Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1614

Dioxin (2,3,7,8-TCDD) in isooctane

NIST SRM 1614

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 6 x 1.2 mL

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in the evaluation of analytical methods in the determination of 2,3,7,8-tetrachlorodibenzo-p-dioxin (2,3,7,8-TCDD). It can also be used to fortify samples with known amounts of 2,3,7,8-TCDD. The 13C-labeled 2,3,7,8-TCDD can be used as an internal standard in methods
based on gas chromatography/mass spectrometry (GC/MS). This SRM consists of separate solutions of unlabeled and labeled 2,3,7,8-tetrachlorodibenzo-p-dioxin (2,3,7,8-TCDD) in 2,4,4-trimethylpentane (isooctane). Three ampoules contain approximately 1.2 mL each of an isooctane solution of unlabeled 2,3,7,8-TCDD, and three
ampoules contain approximately 1.2 mL each of an isooctane solution of 13C-labeled 2,3,7,8-TCDD.

For more information, please refer to the SDS and COA.

SRM 1614_cert SRM 1614 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service