Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1597A

Complex mixture of polycyclic aromatic hydrocarbons coal tar

NIST® SRM® 1597a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 1 x 1.3 mL

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use in the evaluation and validation of analytical methods for the determination of a natural, combustion-related mixture of polycyclic aromatic hydrocarbons (PAHs). SRM 1597a is isolated from a coal tar sample and dissolved in toluene. It is suitable for direct analysis (i.e., without sample cleanup or concentration) in the determination of PAHs using analytical techniques such as gas chromatography (GC), liquid chromatography (LC), or gas chromatography/mass spectrometry (GC/MS). This SRM may also be used to evaluate procedures for measurement of mutagenic activity of combustion-related
mixtures of PAHs and related compounds. A unit of SRM 1597a consists of one 5 mL ampoule, containing 1.3 mL of material.

For more information, please refer to the SDS and COA.

SRM 1597A_cert SRM 1597A _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

39.2 °F - closed cup

flash_point_c

4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service