Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1487

Poly(methyl methacrylate)

NIST® SRM® 1487, 6 K narrow molecular weight distribution

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

NIST standard reference material

autoignition temp.

580 °F

packaging

pkg of 2 g

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

single component solution

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended for use in calibration and performance evaluation of instruments used in polymer technology and science for the determination of molecular weight and molecular weight distribution. It can also be used as a characterized sample for measurements of the limiting viscosity number of poly (methyl methacrylate). The SRM is in the form of a powder. For more information, please refer to the COA and SDS.

SRM 1487_cert SRM 1487 _SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

11 - Combustible Solids

wgk_germany

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service