Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1483A

Linear Polyethylene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Product NIST1483A is currently not available for purchase. Contact Technical Service

grade

certified reference material

packaging

bottle of 10 g

manufacturer/tradename

NIST®

application(s)

chemicals and industrial polymers

format

neat

General description

This Standard Reference Material (SRM) is intended for use in calibration and performance evaluation of instruments used to determine the molecular mass and molecular mass distribution by high temperature size exclusion chromatography (SEC) and instruments
used to obtain the high temperature dilute solution viscosity of the polymer. The product is supplied in the form of a white powder in units of 0.3g.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service