Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST141D

Acetanilide

NIST® SRM® 141d

Synonym(s):

N-Phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H5
CAS Number:
Molecular Weight:
135.16
Beilstein/REAXYS Number:
606468
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

vapor density

4.65 (vs air)

vapor pressure

1 mmHg ( 114 °C)

form

powder

autoignition temp.

1004 °F

packaging

pkg of 2 g

bp

304 °C (lit.)

mp

113-115 °C (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

CC(=O)Nc1ccccc1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

FZERHIULMFGESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

321.8 °F - closed cup

flash_point_c

161 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Is it really organic??Multi-isotopic analysis as a tool to discriminate between organic and conventional plants.
Laurson H K, et al.
Food Chemistry, 141(3), 2812-2820 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service