Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1359B

Coating thickness standard (nonmagnetic coating on steel)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of set (5)

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is designed for calibrating coating thickness gauges that employ magnetic principles. These gauges are used to measure the thickness of nonmagnetic coatings on steel including paint and other organic coatings, as well as nonmagnetic metallic coatings. A unit of SRM 1359b consists of a preconfigured set of five 45 mm × 45 mm coupons: a bare substrate, and four coupons with metallic coatings certified for total coating thickness (nominal coating thicknesses of 48 μm, 140 μm, 505 μm, and 800 μm).This SRM is one in a series of SRMs prepared to cover a coating thickness range from 6 μm to 1935 μm. For more information, please refer to the SDS and COA.

SRM 1359B_cert

SRM 1359B_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service