Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1091A

Oxygen in stainless steel (AISI 431)

NIST SRM 1091a

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$845.00

$845.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 UNIT
$845.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$845.00


Check Cart for Availability

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

grade

certified reference material

Quality Level

packaging

pkg of rod

manufacturer/tradename

NIST®

format

matrix material

General description

Other Notes

Certified for the analytes listed below.
Oxygen (O)

Reference/Informational value is provided for the following analytes. Nitrogen (N)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service