Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST101G

Stainless Steel

NIST® SRM® 101g, 18 Cr - 10 Ni (AISI 304L)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 100 g

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is in the form of a powder and is intended for use in methods of chemical analysis. For more information, please refer to the SDS and COA.

SRM 101G_cert

SRM 101G_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service