Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST1018B

Glass (particle size)

NIST® SRM® 1018b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

beads

packaging

pkg of 87 g

manufacturer/tradename

NIST®

application(s)

pharmaceutical (small molecule)

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating and calibrating particle size measurement instrumentation covering 220 μm to 70 μm range. Typical use is in the evaluation of wire-cloth test sieives in the range from No. 60 (250 μm) through No. 25 (710 μm). The SRM consist of a single bottle containing approximately 87 g of sold spherical soda-lime glass beads. For more information, please refer to the SDS and COA.

SRM 1018B_cert

SRM 1018B_SDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service