Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N15000

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 40049 mPa.s (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

40049 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified is specifically designed, tested and certified for the calibration, verification and certification of glass capillary viscometers and other viscosity measuring equipment where the precise control of temperature is required. ISO 17025 is recognized, internationally for competence to carry out testing and or calibrations to the standard dictated by international methods. This requires accredited facilities to comply with all the requirements detailed within the varied sections of the standard. The highest level of quality assurance is achieved through ISO 17034. This confirms the expertise of the manufacturer, the integrity of the production systems and the reliability of the certified reference materials produced.

Features and Benefits

Manufactured in accordance with ASTM D2162 (Standard Practice for basic calibration of master viscometers and viscosity standard oils).

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service