Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N0559000

Netilmicin sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Netilmicin sulfate salt, 1-N-Ethylsisomicin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H41N5O7 · 5/2H2SO4
CAS Number:
Molecular Weight:
720.78
MDL number:
UNSPSC Code:
12352200

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

InChI

1S/2C21H41N5O7.5H2O4S/c2*1-4-26-13-7-12(24)16(32-19-11(23)6-5-10(8-22)31-19)14(27)17(13)33-20-15(28)18(25-3)21(2,29)9-30-20;5*1-5(2,3)4/h2*5,11-20,25-29H,4,6-9,22-24H2,1-3H3;5*(H2,1,2,3,4)/t2*11-,12+,13-,14+,15-,16-,17+,18-,19-,20-,21+;;;;;/m11...../s1

InChI key

AGFWIZQEWFGATK-UNZHCMSXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Netilmicin sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Craig et al.
Pharmacotherapy, 3(6), 305-315 (1983-11-01)
Netilmicin, the 1-N-ethyl derivative of sisomicin, is a new aminoglycoside antibiotic that was recently marketed in the United States. Its role in therapeutics is not yet established. The pharmacokinetic profile of netilmicin is very similar to that of gentamicin. Its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service