Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSS001

Supelco

Stock Multi-Ion Std 1

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC MSS-001

concentration

in H2O

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Stock Multi-Ion Std 1 may be used as a reference standard for the determination of nitrite and nitrate ions in food products and vegetables using ion chromatography (IC).[1]

Analysis Note

This is a Multi-Ion Solution Standard and contains:
Bromide - 100μg/mL
Chloride - 100μg/mL
Fluoride - 100μg/mL
Nitrate as NO3 - 100μg/mL
Sulfate - 100μg/mL
Phosphate as PO4 - 100μg/mL

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ion chromatographic determination of nitrate and nitrite in vegetable and fruit baby foods
Mcmullen.ES, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 88, 1793-1796 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service