Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MMW001-250ML

Matrix Modifier, Waste Water - Simulated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101600

packaging

pkg of 1 × 250 mL

concentration

in H2O

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Preparation Note

Waste Water - Simulated: A 250 mL sample.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Analyte

Description

    Non-Filterable Residue (TSS) 25.0 +/-0.128
    Turbidity 10.0 +/-0.0511

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service