Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MMS-001-5X20ML

Matrix Modifier, Salt Water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101600
Pricing and availability is not currently available.

packaging

kit of 5 × 20 mL

concentration

in H2O

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Preparation Note

Salt Water: 5 Concentrates for dilution to 10L.

Other Notes

Proficiency testing samples are presented as single blind. No values for the dsamples are released until the testing period concludes. Values of analytes vary lot to lot.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service