MML-001-5X20ML
Matrix Modifier, Moderately Hard Water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
47101600
packaging
kit of 5 × 20 mL
manufacturer/tradename
RTC MML-001K
concentration
in H2O
Looking for similar products? Visit Product Comparison Guide
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Preparation Note
Moderately Hard Water: 5 concentrates for dilution to 10L
Other Notes
Proficiency testing samples are presented as single blind. No values for the samples are released until the testing periiod concludes. Values of analytes vary lot to lot.
Analyte
Description
Hardness, total as CaCO3 100 +/-0.511
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service