Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIC026

Supelco

Salmonella in Soil - P/A 5 Samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Product MIC026 is not currently sold in your country. Contact Technical Service

packaging

set of 1 × 5 ea

manufacturer/tradename

RTC MIC-026

storage temp.

2-8°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO Guide 34.
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

Allow the capped sample vial to equilibrate to room temperature. This sample set contains 5 vials each containing 1g of sludge. Each sample vial is uniquely marked with a sample number. The bacteria are provided in dehydrated form for enhanced stability. They must be re-hydrated prior to performing the tests. It is important that the instructions are followed explicitly. Contact Technical Support if a deviation is required.

Sample preparation
Allow the capped sample vial to equilibrate to room temperature.
Transfer entire contents of vial to blender jar, add the required amount of dilution water.
Blend for 1 to 2 minutes at low speed.
Prepare the appropriate decimal dilutions of the homogenized slurry as quickly as possible to minimize settling.
Assume 1.0g as your sampling weight.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Analyte

Description

    Salmonella -Presence/Absence 0.00 - 1.00

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service