Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIC018

Supelco

Staphylococcus aureus - WS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

packaging

set of 1 × 2 ea

manufacturer/tradename

RTC MIC-018

storage temp.

-10 to -25°C

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Preparation Note

This PT sample is designed for the quantitative analysis of S. aureus. The PT sample is provided in duplicate as a stable dehydrated Vitroid requiring hydration. The additional sample is provided for mishaps in the lab. A 100 mL of sterile hydration fluid is supplied with the PT sample. This is a phosphate buffer. The dehydrated Vitroid sample is stable for a time exceeding the study schedule.

Sample preparation

Allow the capped sample vial and hydrating fluid to equilibrate to room temperature.
The hydration fluid is sterile and may be stored at room temperature up to the expiration date listed on the bottom, refrigeration is not necessary for the hydration fluid.
Open the sample vial and aseptically transfer the Vitroid to the hydrating fluid.
NOTE: The Vitroid is the colored disk on top of the cotton. The opaque beads below the cotton keep container dry and are not biologicals.
Allow the Vitroid to dissolve completely this may take up to 15 minutes.
Mix by inverting gently several times, shaking may cause the cap to leak. This represents the sample for analysis. Incubate at the temperature required by your method.
Temperature requirements are rigorous, higher temperatures will kill the organisms and lower temperatures may suppress growth.
Minimum incubation time is 24 hours. Further may be necessary depending on the method and media.
Further dilutions are recommended to achieve a proper count, but base all counts on the whole volume sample.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Legal Information

Analyte

Description

    Staphylococcus aureus 0.000 - 10,000 CFU

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service