Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MIC013

Supelco

Salmonella PT- Sludge

Proficiency Testing Material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.85

CofA

current certificate can be downloaded

packaging

set of 2 ea

manufacturer/tradename

RTC MIC-013

application(s)

environmental
food and beverages

shipped in

wet ice

storage temp.

−20°C

suitability

Salmonella spp.

General description

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Packaging

Set contains 2 sterile screw cap vials each containing a Vitroid (a small disk containing the possible organisms) and a 50 g container of sterile sludge.

Other Notes

Proficiency testing samples are presented as single blind. No values for the samples are released until the testing period concludes. Values of analytes vary lot to lot.

Proficiency Testing Schedule (2019-2020)

Soils
LPTP19-S1January 30 - March 15, 2019
LPTP19-S2April 10 - May 24, 2019
LPTP19-S3 July 24 - September 6, 2019
LPTP19-S4 October 9 - November 22, 2019

LPTP20-S1January 29 - March 13, 2020
LPTP20-S2April 8 - May 22, 2020
LPTP20-S3 July 22 - September 4, 2020
LPTP20-S4 October 7 - November 20, 2020

Quick-Turn Study - (non-scheduled proficiency test that is available at any time)
This PT sample is designed for the quantitative analysis of Salmonella. The PT sample is provided in duplicate as a stable dehydrated vitroid requiring hydration. The additional sample is provided for mishaps in the lab. Store the PT sample as directed. The dehydrated vitroid sample is stable for a time exceeding the study schedule.

Study Schedule

Legal Information

Analyte

Description

    Salmonella 1.00 - 1000 MPN/100 mL

related product

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service