form
liquid
shipped in
wet ice
storage temp.
2-8°C
General description
The supplement is for use with human hepatocyte platting medium to prepare required volumes of complete media. Upon receiving a shipment of supplement it is important the end user gives the shipment attention without delay. To ensure the highest quality of the supplement, upon receipt supplement should be immediately stored at –20 °C.
Application
For use with Human Hepatocyte Culture Medium. See technical bulletin for LifeNet Health® Primary Human Hepatocytes for protocols.
Legal Information
LifeNet Health is a registered trademark of LifeNet Health
required but not provided
Product No.
Description
Pricing
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service