Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8402

Supelco

Mefexamide hydrochloride

analytical standard, for drug analysis

Synonym(s):

p-Methoxyphenoxyacetic acid 2-(diethylamino)ethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O3 · HCl
CAS Number:
Molecular Weight:
316.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.CCN(CC)CCNC(=O)COc1ccc(OC)cc1

InChI

1S/C15H24N2O3.ClH/c1-4-17(5-2)11-10-16-15(18)12-20-14-8-6-13(19-3)7-9-14;/h6-9H,4-5,10-12H2,1-3H3,(H,16,18);1H

InChI key

AXBHPHPHZHAOAK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Preliminary pharmacological investigations of six new p-chlorphenoxyacetic acid derivatives (author's transl)].
A Kruszewska et al.
Annales Universitatis Mariae Curie-Sklodowska. Sectio D: Medicina, 34, 259-268 (1979-01-01)
[Correction of psychodepressive effects of benzodiazepine tranquilizers by administration of psychostimulants].
Iu G Bobkov et al.
Biulleten' eksperimental'noi biologii i meditsiny, 94(10), 48-51 (1982-10-01)
W Gielsdorf et al.
Zeitschrift fur Rechtsmedizin. Journal of legal medicine, 87(4), 297-303 (1981-01-01)
After oral ingestion of 400 mg Mefexamidehydrochloride for Mefexamide (I) and its main degradation product Desmethyl-Mefexamide (II) the following pharmakokinetic parameters have been determined: 1. Elimination of I and II follows 1st order kinetics. 2. Biological half-life t 1/2 for
W Gielsdorf
Zeitschrift fur Rechtsmedizin. Journal of legal medicine, 87(1-2), 117-127 (1981-01-01)
After oral administration of a therapeutic dosage (400 mg) Mefexamide (I), six excretion products were detected in human urine and subsequently identified by GC/MS and TLC: in addition to the unchanged drug (I) in the acidic extracted urine without hydrolysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service