Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8163

Supelco

Leucomisine

analytical standard, 98% (TLC), amorphous powder

Synonym(s):

Desacetoxymatricarin, Leucodine, Leucomisin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18O3
Molecular Weight:
246.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

98% (TLC)

form

amorphous powder

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

Leucomisine is a natural sequiterpene lactone of the guaiane type, isolated from the essential oil of Artemisia ludoviciana.

Application

Leucomisine may be used as an analytical reference standard for the quantification of the analyte in the essential oil of Artemisia ludoviciana using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Antiatherosclerotic, angioprotective, hypolipidemic and anti-inflammatory.

Warning

Protect from light.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The investigation of natural compounds by HPLC. III. A study of sesquiterpene lactones of the guaiane type by microcolumn HPLC
Nuriddinova.R.M, et al.
Chemistry of Natural Compounds, 29(4), 458-461 (1993)
Composition of the essential oil and of some extracts of the aerial parts of Artemisia ludoviciana var. latiloba Nutt
Collin G, et al.
Analytical Sciences, 19(2), 28-38 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service