Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8038

Supelco

Lapiferine

analytical standard, 97% (TLC), solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H34O6
Molecular Weight:
394.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

97% (TLC)

form

solid

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C\C=C/C(=O)O[C@@H]1C[C@@H]2O[C@@H]2[C@H](O)[C@]3(C)CC[C@](O)(C(C)C)[C@@]13C

InChI

1S/C19H30O5/c1-6-7-14(20)24-13-10-12-15(23-12)16(21)17(4)8-9-19(22,11(2)3)18(13,17)5/h6-7,11-13,15-16,21-22H,8-10H2,1-5H3/b7-6-/t12-,13+,15-,16-,17-,18-,19-/m0/s1

InChI key

YNHWIAXMARBYFN-WNFXZRHOSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Warning

Protect from light.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service