Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L0100000

Lactose

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-Lactose monohydrate, β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Beilstein/REAXYS Number:
3768231
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

API family

lactose

analyte chemical class(es)

oligosaccharides

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

HBDJFVFTHLOSDW-XBLONOLSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lactose EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel M Martinez et al.
Journal of clinical microbiology, 52(7), 2521-2529 (2014-05-09)
The identification of organisms from positive blood cultures generally takes several days. However, recently developed rapid diagnostic methods offer the potential for organism identification within only a few hours of blood culture positivity. In this study, we evaluated the performance
Muireann Egan et al.
Applied and environmental microbiology, 80(14), 4414-4426 (2014-05-13)
Bifidobacteria constitute a specific group of commensal bacteria that inhabit the gastrointestinal tracts of humans and other mammals. Bifidobacterium breve UCC2003 has previously been shown to utilize several plant-derived carbohydrates that include cellodextrins, starch, and galactan. In the present study
Xiaoge Gao et al.
Oncotarget, 5(8), 2077-2084 (2014-05-09)
The presence and level of circulating galectin-3 (Gal-3), a member of the galectin family, is associated with diverse diseases ranging from heart failure, immune disorders to cancer metastasis and serves as a biomarker of diagnosis and treatment response. However, the
Srimanta Sarkar et al.
International journal of pharmaceutics, 475(1-2), 558-565 (2014-09-15)
This study investigated the influence of different disintegrants, present in different substrate physical forms, on dimensional recovery of multi-component tablets prepared at different compression pressures. Formulations containing model drug, metformin, (10%, w/w), different disintegrants (10%, w/w), and lactose (80%, w/w)
Song Guo et al.
Brain : a journal of neurology, 137(Pt 11), 2951-2959 (2014-08-28)
The initiating mechanisms of migraine attacks are very complex but may involve the cyclic AMP signalling pathway. It is unknown whether intracellular cyclic AMP accumulation induces migraine attacks. We investigated whether administration of cilostazol, which causes cyclic AMP accumulation, may

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service