Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM804

Rice flour (trace elements)

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

format

matrix material

storage temp.

2-8°C

General description

Rice flour (trace elements) has been used as a reference material in the analysis of minerals in highly consumed cereal product samples using atomic absorption spectrophotometry (AAS).[1]

Analysis Note

For more information please see:
IRMM804

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements: Arsenic (As), Cadmium (Cd), Copper (Cu), Lead (Pb), Manganese (Mn), Selenium (Se), Zinc (Zn)

Matrix Group: Cereals, Bread, Rice and Maize

Legal Information

IRMM is a registered trademark of European Commission

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Phytate, zinc, iron and calcium content of common Bolivian food, and implications for mineral bioavailability
Lazarte.EC, et al.
J. Food Compos. Anal., 39, 111-119 (2015)
Dandi Hou et al.
Applied and environmental microbiology, 84(12) (2018-04-15)
Rhizospheric bacteria play important roles in plant tolerance and activation of heavy metals. Understanding the bacterial rhizobiome of hyperaccumulators may contribute to the development of optimized phytoextraction for metal-polluted soils. We used 16S rRNA gene amplicon sequencing to investigate the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service