Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM801

Triglycerides in cocoa butter

IRMM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

shipped in

dry ice

storage temp.

−20°C

General description

Certified for the relative mass fractions of the triglycerides POP, POS, POO, SOS, SOO

Application

Triglycerides in cocoa butter has been used as a standard reference material to determine the concentration of high di-saturated triacylglycerol (TAG) formed by the lipase catalyzed acidolysis of high oleic sunflower oil with a combination of stearic–palmitic acid using GC.[1]

Analysis Note

For more information please see:
IRMM801

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Lipids and Fatty acids: 1,2-Dioleoyl-3-palmitoyl-glycerol, 1,2-Dioleoyl-3-stearoyl-glycerol, 1,3-Dipalmitoyl-2-oleoylglycerol, 1,3-Distearoyl-2-oleoylglycerol, 1-Palmitoyl-2-oleoyl-3-stearoyl-glycerol

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

IRMM is a registered trademark of European Commission

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic study of the acidolysis of high oleic sunflower oil with stearic?palmitic acid mixtures catalysed by immobilised Rhizopus oryzae lipase
Ray J, et al.
Biochemical Engineering Journal, 73, 17-28 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service