Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM634

Iron-56Fe solution

IRMM®, certified reference material, 1.8 M in hydrochloric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

agency

IRMM®

manufacturer/tradename

JRC

concentration

1.8 M in hydrochloric acid

application(s)

pharmaceutical (small molecule)

format

single component solution

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM634

Legal Information

IRMM is a registered trademark of European Commission

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service