Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IRMM619

Rubidium-85Rb solution

IRMM®, certified reference material, 0.5 M in nitric acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$1,010.00

$1,010.00


Estimated to ship onJuly 18, 2025FromSAINT LOUIS


Request a Bulk Order

Select a Size

Change View
5 ML
$1,010.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,010.00


Estimated to ship onJuly 18, 2025FromSAINT LOUIS


Request a Bulk Order

grade

certified reference material

agency

IRMM®

isotopic purity

 85Rb

manufacturer/tradename

JRC

concentration

0.5 M in nitric acid

application(s)

general analytical

format

single component solution

General description

Isotope abundance ratio

Analysis Note

For more information please see:
IRMM619

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service